missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Diiodo-trans-stilène 97,0+%, TCI America™
Spécifications
Spécifications
| Quantity | 5 g |
| Formula Weight | 432.04 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | 4,4′-Diiodo-trans-stilbene |
Sécurité et manipulation
| TSCA | No |