missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diiodo-trans-stilbene 97.0+%, TCI America™

Catalog No. D30941G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D30941G 1 g
D30945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D30941G Supplier TCI America Supplier No. D30941G

Chemical Identifiers

CAS 201861-91-8
Molecular Formula C14H10I2
Molecular Weight (g/mol) 432.043
InChI Key BYEYAZGFUKIZPM-OWOJBTEDSA-N
PubChem CID 21709796
IUPAC Name 1-iodo-4-[(E)-2-(4-iodophenyl)ethenyl]benzene
SMILES C1=CC(=CC=C1C=CC2=CC=C(C=C2)I)I

Specifications

Quantity 1 g
Formula Weight 432.04
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diiodo-trans-stilbene