missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dihexyl-2,2'-bithiophene 95.0+%, TCI America™

Catalog No. D4182200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4182200MG 200 mg
D41821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4182200MG Supplier TCI America Supplier No. D4182200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135926-94-2
Molecular Formula C20H30S2
Molecular Weight (g/mol) 334.58
InChI Key RYQPWKFBXWPBGB-UHFFFAOYSA-N
PubChem CID 15540627
IUPAC Name 4-hexyl-2-(4-hexylthiophen-2-yl)thiophene
SMILES CCCCCCC1=CSC(=C1)C2=CC(=CS2)CCCCCC

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 334.58
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4′-Dihexyl-2,2′-bithiophene
TSCA No
Recommended Storage Refrigerator