missing translation for 'onlineSavingsMsg'
Learn More

4,4-Difluoropiperidine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. p-7047645
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1955503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1955503 Supplier Thermo Scientific Chemicals Supplier No. L1955503

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144230-52-4
Molecular Formula C5H10ClF2N
Molecular Weight (g/mol) 157.589
MDL Number MFCD03095381
InChI Key OABUKBBBSMNNPM-UHFFFAOYSA-N
Synonym 4,4-difluoropiperidine hydrochloride, 4,4-difluoropiperidine hcl, piperidine, 4,4-difluoro-, hydrochloride, 4,4-difluoropiperidine, chloride, 4,4-difluoropiperidin-1-ium chloride, 4,4-difluoropiperidinehydrochloride, pubchem6797, acmc-1bwk7, 4,4-difluoropiperidinehcl, 4,4-difluoropiperidin hcl
PubChem CID 2758351
IUPAC Name 4,4-difluoropiperidine;hydrochloride
SMILES C1CNCCC1(F)F.Cl

Specifications

Melting Point ∼172°C
Quantity 1 g
Beilstein 6812163
Sensitivity Moisture sensitive
Formula Weight 157.59
Percent Purity 98%
Chemical Name or Material 4,4-Difluoropiperidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only