missing translation for 'onlineSavingsMsg'
Learn More

4,4-Difluorocyclohexanone, 97%, Thermo Scientific Chemicals

Catalog No. p-7028633
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26027MD 250 mg
AAH2602703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH26027MD Supplier Thermo Scientific Chemicals Supplier No. H26027MD

4,4-Difluorocyclohexanone is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4-Difluorocyclohexanone is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 22515-18-0
Molecular Formula C6H8F2O
Molecular Weight (g/mol) 134.126
MDL Number MFCD08146635
InChI Key NYYSPVRERVXMLJ-UHFFFAOYSA-N
Synonym 4,4-difluorocyclohexanone, 4,4-difluoro-cyclohexanone, cyclohexanone, 4,4-difluoro, difluorocyclohexanone, acmc-1cnv9, 4,4-difluoro cyclohexanone, 4,4-difluoro-1-cyclohexanone, 4,4-difluorocycloxanone, 1,1,-difluoro-4-oxocyclohexane
PubChem CID 15789092
IUPAC Name 4,4-difluorocyclohexan-1-one
SMILES C1CC(CCC1=O)(F)F

Specifications

Melting Point 34°C to 40°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 134.12
Percent Purity 97%
Chemical Name or Material 4,4-Difluorocyclohexanone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only