missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Difluorocyclohexanecarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America D43305G
Specifications
| 4,4-Difluorocyclohexanecarboxylic Acid | |
| 107°C | |
| C7H10F2O2 | |
| 5 g | |
| HYIUDFLDFSIXTR-UHFFFAOYSA-N | |
| 4,4-difluorocyclohexane-1-carboxylic acid | |
| 2779136 | |
| ≥98.0% (GC,T) |
| 122665-97-8 | |
| White | |
| MFCD03788493 | |
| 4,4-difluorocyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4,4-difluoro, 4,4-difluorocyclohexylcarboxylic acid, 4,4-difluorocyclohexanoic acid, 4, 4-difluoro-cyclohexane carboxylic acid, 4,4-difluorocyclohexane carboxylic acid, 4,4-difluorocyclohexanecarboxylicacid, pubchem15915, acmc-209aln, ksc174k3t | |
| C1CC(CCC1C(=O)O)(F)F | |
| 164.152 | |
| 164.15 | |
| Crystalline Powder |
Chemical Identifiers
| 122665-97-8 | |
| 164.152 | |
| HYIUDFLDFSIXTR-UHFFFAOYSA-N | |
| 2779136 | |
| C1CC(CCC1C(=O)O)(F)F |
| C7H10F2O2 | |
| MFCD03788493 | |
| 4,4-difluorocyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4,4-difluoro, 4,4-difluorocyclohexylcarboxylic acid, 4,4-difluorocyclohexanoic acid, 4, 4-difluoro-cyclohexane carboxylic acid, 4,4-difluorocyclohexane carboxylic acid, 4,4-difluorocyclohexanecarboxylicacid, pubchem15915, acmc-209aln, ksc174k3t | |
| 4,4-difluorocyclohexane-1-carboxylic acid |
Safety and Handling
TSCA : No