missing translation for 'onlineSavingsMsg'
Learn More

4,4-Difluorocyclohexanecarboxylic Acid 98.0+%, TCI America™

Catalog No. D43301G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43301G 1 g
D43305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43301G Supplier TCI America Supplier No. D43301G

Chemical Identifiers

CAS 122665-97-8
Molecular Formula C7H10F2O2
Molecular Weight (g/mol) 164.152
MDL Number MFCD03788493
InChI Key HYIUDFLDFSIXTR-UHFFFAOYSA-N
Synonym 4,4-difluorocyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4,4-difluoro, 4,4-difluorocyclohexylcarboxylic acid, 4,4-difluorocyclohexanoic acid, 4, 4-difluoro-cyclohexane carboxylic acid, 4,4-difluorocyclohexane carboxylic acid, 4,4-difluorocyclohexanecarboxylicacid, pubchem15915, acmc-209aln, ksc174k3t
PubChem CID 2779136
IUPAC Name 4,4-difluorocyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)(F)F

Specifications

Melting Point 107°C
Color White
Quantity 1 g
Formula Weight 164.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4-Difluorocyclohexanecarboxylic Acid