missing translation for 'onlineSavingsMsg'
Learn More

4,4-Difluorocyclohexanecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog No. p-7028490
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2583103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2583103 Supplier Thermo Scientific Chemicals Supplier No. H2583103

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122665-97-8
Molecular Formula C7H10F2O2
Molecular Weight (g/mol) 164.152
MDL Number MFCD03788493
InChI Key HYIUDFLDFSIXTR-UHFFFAOYSA-N
Synonym 4,4-difluorocyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4,4-difluoro, 4,4-difluorocyclohexylcarboxylic acid, 4,4-difluorocyclohexanoic acid, 4, 4-difluoro-cyclohexane carboxylic acid, 4,4-difluorocyclohexane carboxylic acid, 4,4-difluorocyclohexanecarboxylicacid, pubchem15915, acmc-209aln, ksc174k3t
PubChem CID 2779136
IUPAC Name 4,4-difluorocyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)(F)F

Specifications

Melting Point 104°C to 107°C
Quantity 1 g
Formula Weight 164.15
Percent Purity 98%
Chemical Name or Material 4,4-Difluorocyclohexanecarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only