missing translation for 'onlineSavingsMsg'
Learn More

4,4-Diethylamino-2-butyn-1-ol, 98%

Catalog No. p-7035774
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5350106 5 g
AAH5350114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5350106 Supplier Thermo Scientific Chemicals Supplier No. H5350106
Only null left

CAS: 10575-25-4 | C8H15NO | 141.21 g/mol

4,4-Diethylamino-2-butyn-1-ol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4-Diethylamino-2-butyn-1-ol is used as pharmaceutical intermediate.

Solubility
Soluble in chloroform and ethyl acetate. Soluble in water 11 g/L @25°C.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 10575-25-4
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.21
MDL Number MFCD00671356
InChI Key ACGZBRWTWOZSFU-UHFFFAOYSA-N
PubChem CID 82735
IUPAC Name 4-(diethylamino)but-2-yn-1-ol
SMILES CCN(CC)CC#CCO

Specifications

Density 0.952
Boiling Point 231°C to 232°C
Flash Point 104°C (219°F)
Refractive Index 1.479
Linear Formula (CH3CH2)2NCH2C≡CCH2OH
Quantity 5 g
Synonym 4-diethylamino but-2-yn-1-ol, 4-diethylamino-2-butyn-1-ol, unii-ma2de62u5w, ma2de62u5w, 2-butyn-1-ol,4-diethylamino, 2-butyn-1-ol, 4-diethylamino, acmc-2098hk, 4-diethylamino-2-butyne-ol, 1-diethylaminobut-2-yn-4-ol
Solubility Information Soluble in chloroform and ethyl acetate. Soluble in water 11g/L @25°C.
Formula Weight 141.21
Percent Purity 98%
Chemical Name or Material 4,4-Diethylamino-2-butyn-1-ol
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 234-163-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only