missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Diethoxybutylamine 98.0+%, TCI America™
Supplier: TCI America D396825G
Specifications
| 4,4-Diethoxybutylamine | |
| Yellow | |
| C8H19NO2 | |
| 25 g | |
| 4,4-diethoxybutylamine, 4-aminobutyraldehyde diethyl acetal, 1-butanamine, 4,4-diethoxy, 4,4-diethoxy-1-butanamine, ambda, 4,4-diethoxy-butylamine, acmc-2097jf, 4-aminobutanal diethyl acetal, ksc356g0p, 4,4-diethoxy-1-butanamine # | |
| CCOC(CCCN)OCC | |
| 161.25 | |
| 161.25 | |
| Liquid |
| 6346-09-4 | |
| 196°C | |
| MFCD00008227 | |
| 2735 | |
| GFLPSABXBDCMCN-UHFFFAOYSA-N | |
| 4,4-diethoxybutan-1-amine | |
| 80672 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 6346-09-4 | |
| 161.25 | |
| GFLPSABXBDCMCN-UHFFFAOYSA-N | |
| 80672 | |
| CCOC(CCCN)OCC |
| C8H19NO2 | |
| MFCD00008227 | |
| 4,4-diethoxybutylamine, 4-aminobutyraldehyde diethyl acetal, 1-butanamine, 4,4-diethoxy, 4,4-diethoxy-1-butanamine, ambda, 4,4-diethoxy-butylamine, acmc-2097jf, 4-aminobutanal diethyl acetal, ksc356g0p, 4,4-diethoxy-1-butanamine # | |
| 4,4-diethoxybutan-1-amine |
Safety and Handling
TSCA : No