missing translation for 'onlineSavingsMsg'
Learn More

4,4-Diethoxybutylamine 98.0+%, TCI America™

Catalog No. D39685G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39685G 5 g
D396825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39685G Supplier TCI America Supplier No. D39685G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6346-09-4
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.25
MDL Number MFCD00008227
InChI Key GFLPSABXBDCMCN-UHFFFAOYSA-N
Synonym 4,4-diethoxybutylamine, 4-aminobutyraldehyde diethyl acetal, 1-butanamine, 4,4-diethoxy, 4,4-diethoxy-1-butanamine, ambda, 4,4-diethoxy-butylamine, acmc-2097jf, 4-aminobutanal diethyl acetal, ksc356g0p, 4,4-diethoxy-1-butanamine #
PubChem CID 80672
IUPAC Name 4,4-diethoxybutan-1-amine
SMILES CCOC(CCCN)OCC

Specifications

Color Yellow
Boiling Point 196°C
Quantity 5 g
UN Number 2735
Formula Weight 161.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4,4-Diethoxybutylamine