missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorochalcone 98.0+%, TCI America™

Catalog No. D48465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48465G 5 g
D484625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48465G Supplier TCI America Supplier No. D48465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19672-59-4
Molecular Formula C15H10Cl2O
Molecular Weight (g/mol) 277.14
MDL Number MFCD00018704
InChI Key YMEMCRBNZSLQCQ-XCVCLJGOSA-N
Synonym 4,4'-dichlorochalcone, e-1,3-bis-4-chloro-phenyl-propenone, e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl-2-propen-1-one, 4',4-dichlorochalcone, e-4,4'-dichlorochalcone, trans-1,3-di-4-chlorophenyl-prop-2-en-1-one
PubChem CID 5377011
IUPAC Name (2E)-1,3-bis(4-chlorophenyl)prop-2-en-1-one
SMILES ClC1=CC=C(\C=C\C(=O)C2=CC=C(Cl)C=C2)C=C1

Specifications

Melting Point 158°C
Color Yellow
Quantity 5 g
Formula Weight 277.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dichlorochalcone