missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorochalcone, 98+%

Catalog No. AAL1071706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1071706 5 g
AAL1071714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1071706 Supplier Thermo Scientific Chemicals Supplier No. L1071706

CAS: 19672-59-4 | C15H10Cl2O | 277.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19672-59-4
Molecular Formula C15H10Cl2O
Molecular Weight (g/mol) 277.14
MDL Number MFCD00018704
InChI Key YMEMCRBNZSLQCQ-XCVCLJGOSA-N
Synonym 4,4'-dichlorochalcone, e-1,3-bis-4-chloro-phenyl-propenone, e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl-2-propen-1-one, 4',4-dichlorochalcone, e-4,4'-dichlorochalcone, trans-1,3-di-4-chlorophenyl-prop-2-en-1-one
PubChem CID 5377011
IUPAC Name (E)-1,3-bis(4-chlorophenyl)prop-2-en-1-one
SMILES ClC1=CC=C(\C=C\C(=O)C2=CC=C(Cl)C=C2)C=C1

Specifications

Melting Point 156°C to 160°C
Quantity 5 g
Beilstein 2331589
Formula Weight 277.15
Percent Purity ≥98%
Chemical Name or Material 4,4'-Dichlorochalcone
RTECSNumber UD5572635
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only