missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dicarboxy-2,2'-bipyridine, 98%

Catalog No. AC363660010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC363660010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC363660010 Supplier Thermo Scientific Chemicals Supplier No. 363660010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6813-38-3 | C12H6N2O4 | 242.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6813-38-3
Molecular Formula C12H6N2O4
Molecular Weight (g/mol) 242.19
MDL Number MFCD00015430
InChI Key FXPLCAKVOYHAJA-UHFFFAOYSA-L
Synonym 2,2'-bipyridine-4,4'-dicarboxylic acid, 2,2'-bipyridyl-4,4'-dicarboxylic acid, 4,4'-dicarboxy-2,2'-bipyridine, 2,2'-biisonicotinic acid, 2,2'-dipyridyl-4,4'-dicarboxylic acid, 2,2'-bipyridine-4,4'-dicarboxylicacid, 2-4-carboxypyridin-2-yl pyridine-4-carboxylic acid, 4,4-dicarboxy-2,2-bipyridine, 2-4-carboxy-2-pyridyl pyridine-4-carboxylic acid
PubChem CID 688094
IUPAC Name 2-(4-carboxypyridin-2-yl)pyridine-4-carboxylic acid
SMILES [O-]C(=O)C1=CC=NC(=C1)C1=CC(=CC=N1)C([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Beilstein 25, V,6, 36
Solubility Information Solubility in water: insoluble in water
Formula Weight 244.21
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4, 4'-Dicarboxy-2, 2'-bipyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only