missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromobenzophenone, 98+%

Catalog No. AAL0820006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0820006 5 g
AAL0820003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0820006 Supplier Thermo Scientific Chemicals Supplier No. L0820006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3988-03-2 | C5H10O2 | 102.13 g/mol

4,4'-Dibromobenzophenone is used in the synthesis of high molecular weight poly(p-phenylene) derivatives through palladium-catalyzed polycondensation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dibromobenzophenone is used in the synthesis of high molecular weight poly(p-phenylene) derivatives through palladium-catalyzed polycondensation reaction.

Solubility
Soluble in ethanol, 2-propanol, acetone and toluene. Miscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 3-2-3988

Specifications

Melting Point 172°C to 176°C
Molecular Formula C5H10O2
MDL Number MFCD00085264
Quantity 5 g
Beilstein 2049958
Synonym ethyl glycidyl ether, 2-ethoxymethyl oxirane, oxirane, ethoxymethyl, 1,2-epoxy-3-ethoxypropane, ethoxymethyl oxirane, 3-ethoxy-1,2-epoxypropane, oxirane, 2-ethoxymethyl, propane, 1,2-epoxy-3-ethoxy, 1-ethoxy-2,3-epoxypropane, ccris 1018
Solubility Information Soluble in ethanol,2-propanol,acetone and toluene. Miscible with water.
InChI Key HQCSZRIVJVOYSU-UHFFFAOYNA-N
SMILES CCOCC1CO1
IUPAC Name 2-(ethoxymethyl)oxirane
Molecular Weight (g/mol) 102.13
PubChem CID 19919
Formula Weight 340.03
Percent Purity ≥98%
Chemical Name or Material 4,4'-Dibromobenzophenone
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-632-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only