missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dibromobenzil 97.0+%, TCI America™
$223.68 - $562.95
Chemical Identifiers
| CAS | 35578-47-3 |
|---|---|
| Molecular Formula | C14H8Br2O2 |
| Molecular Weight (g/mol) | 368.024 |
| MDL Number | MFCD00000104 |
| InChI Key | NYCBYBDDECLFPE-UHFFFAOYSA-N |
| Synonym | 4,4'-dibromobenzil, ethanedione, bis 4-bromophenyl, p,p'-dibromobenzil, 1,2-bis 4-bromophenyl ethane-1,2-dione, benzyl, 4,4'-dibromo, unii-o4txv8a656, bis 4-bromophenyl ethanedione, 1,2-ethanedione, 1,2-bis 4-bromophenyl, bis 4-bromophenyl ethane-1,2-dione, 1,2-bis-4-bromo-phenyl-ethane-1,2-dione |
| PubChem CID | 96430 |
| IUPAC Name | 1,2-bis(4-bromophenyl)ethane-1,2-dione |
| SMILES | C1=CC(=CC=C1C(=O)C(=O)C2=CC=C(C=C2)Br)Br |
Chemical Identifiers
| 35578-47-3 | |
| 368.024 | |
| NYCBYBDDECLFPE-UHFFFAOYSA-N | |
| 96430 | |
| C1=CC(=CC=C1C(=O)C(=O)C2=CC=C(C=C2)Br)Br |
| C14H8Br2O2 | |
| MFCD00000104 | |
| 4,4'-dibromobenzil, ethanedione, bis 4-bromophenyl, p,p'-dibromobenzil, 1,2-bis 4-bromophenyl ethane-1,2-dione, benzyl, 4,4'-dibromo, unii-o4txv8a656, bis 4-bromophenyl ethanedione, 1,2-ethanedione, 1,2-bis 4-bromophenyl, bis 4-bromophenyl ethane-1,2-dione, 1,2-bis-4-bromo-phenyl-ethane-1,2-dione | |
| 1,2-bis(4-bromophenyl)ethane-1,2-dione |
Specifications
| 35578-47-3 | |
| C14H8Br2O2 | |
| 5 g | |
| NYCBYBDDECLFPE-UHFFFAOYSA-N | |
| 1,2-bis(4-bromophenyl)ethane-1,2-dione | |
| 96430 | |
| ≥97.0% (GC) | |
| 4,4′-Dibromobenzil |
| 227°C | |
| MFCD00000104 | |
| 4,4'-dibromobenzil, ethanedione, bis 4-bromophenyl, p,p'-dibromobenzil, 1,2-bis 4-bromophenyl ethane-1,2-dione, benzyl, 4,4'-dibromo, unii-o4txv8a656, bis 4-bromophenyl ethanedione, 1,2-ethanedione, 1,2-bis 4-bromophenyl, bis 4-bromophenyl ethane-1,2-dione, 1,2-bis-4-bromo-phenyl-ethane-1,2-dione | |
| C1=CC(=CC=C1C(=O)C(=O)C2=CC=C(C=C2)Br)Br | |
| 368.024 | |
| 368.02 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes