missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromobenzil, 97%

Catalog No. p-7020956
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1309009 10 g
AAA1309018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1309009 Supplier Thermo Scientific Chemicals Supplier No. A1309009
Only null left

CAS: 35578-47-3 | C14H8Br2O2 | 368.024 g/mol

4,4?-Dibromobenzil was used in preparation of 5,5?-bis -(4-bromo-phenyl)-3-methyl-imidazolidine-2,4-dione, 5,5?- bis -(4-bromo-phenyl)-3-butyl-imidazolidine-2,4-dione, 5,5?-bis-(4-bromo-phenyl)-3-pentyl-imidazolidine-2,4-dione and 5,5?-bis -(4-chloro-phenyl)-3-ethyl imidazolidine-2,4-dione. It was also used in synthesis of novel quinoxaline-based sensitizers for dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dibromobenzil was used in preparation of 5,5′-bis -(4-bromo-phenyl)-3-methyl-imidazolidine-2,4-dione, 5,5′- bis -(4-bromo-phenyl)-3-butyl-imidazolidine-2,4-dione, 5,5′-bis-(4-bromo-phenyl)-3-pentyl-imidazolidine-2,4-dione and 5,5′-bis -(4-chloro-phenyl)-3-ethyl imidazolidine-2,4-dione. It was also used in synthesis of novel quinoxaline-based sensitizers for dye-sensitized solar cells.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 35578-47-3
Molecular Formula C14H8Br2O2
MDL Number MFCD00000104
InChI Key NYCBYBDDECLFPE-UHFFFAOYSA-N
PubChem CID 96430

Specifications

Melting Point 224°C to 226°C
Quantity 10 g
Synonym 4,4'-dibromobenzil, ethanedione, bis 4-bromophenyl, p,p'-dibromobenzil, 1,2-bis 4-bromophenyl ethane-1,2-dione, benzyl, 4,4'-dibromo, unii-o4txv8a656, bis 4-bromophenyl ethanedione, 1,2-ethanedione, 1,2-bis 4-bromophenyl, bis 4-bromophenyl ethane-1,2-dione, 1,2-bis-4-bromo-phenyl-ethane-1,2-dione
Solubility Information Sparingly soluble in water.
SMILES C1=CC(=CC=C1C(=O)C(=O)C2=CC=C(C=C2)Br)Br
IUPAC Name 1,2-bis(4-bromophenyl)ethane-1,2-dione
Molecular Weight (g/mol) 368.024
Formula Weight 368.04
Percent Purity 97%
Chemical Name or Material 4,4'-Dibromobenzil
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 252-628-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only