missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromo-2,2'-bis(triisopropylsilyl)-5,5'-bithiazole 98.0+%, TCI America™

Catalog No. d4605200mg
Change view
Click to view available options
Quantity:
200 mg
Catalog No. Quantity
D4605200MG 200 mg
1 options

Catalog No. D4605200MG

Supplier: TCI America D4605200MG

Only null left

Chemical Identifiers

CAS 1223559-98-5
Molecular Formula C24H42Br2N2S2Si2
Molecular Weight (g/mol) 638.71
MDL Number MFCD27923042
InChI Key HNWAYAXELMAWTB-UHFFFAOYSA-N
PubChem CID 66728310
IUPAC Name 4-bromo-5-{4-bromo-2-[tris(propan-2-yl)silyl]-1,3-thiazol-5-yl}-2-[tris(propan-2-yl)silyl]-1,3-thiazole
SMILES CC(C)[Si](C(C)C)(C(C)C)C1=NC(Br)=C(S1)C1=C(Br)N=C(S1)[Si](C(C)C)(C(C)C)C(C)C

Specifications

Melting Point 113°C
Quantity 200 mg
Formula Weight 638.71
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dibromo-2,2′-bis(triisopropylsilyl)-5,5′-bithiazole