missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diaminodiphenylamine Sulfate Hydrate 97.0+%, TCI America™

Catalog No. p-7125999
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D004125G 25 g
D0041500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D004125G Supplier TCI America Supplier No. D004125G

Chemical Identifiers

CAS 53760-27-3
Molecular Formula C12H15N3O4S
Molecular Weight (g/mol) 297.329
MDL Number MFCD00013113
InChI Key OOZQLPDAELLDNY-UHFFFAOYSA-N
Synonym 4,4'-diaminodiphenylamine sulfate, n1-4-aminophenyl benzene-1,4-diamine sulfate, unii-02x5ljd26t, n4-4-aminophenyl benzene-1,4-diamine; sulfuric acid, n1-4-aminophenyl benzene-1,4-diamine; sulfuric acid, 4,4'-iminodianiline sulfate, n-4-aminophenyl benzene-1,4-diamine sulfate 1:1, 4,4'-diaminodiphenylaminsulfat, bis 4-aminophenyl amine sulfate, n-4-aminophenyl benzene-1,4-diamine sulphate 1:1
PubChem CID 104599
IUPAC Name 4-N-(4-aminophenyl)benzene-1,4-diamine;sulfuric acid
SMILES C1=CC(=CC=C1N)NC2=CC=C(C=C2)N.OS(=O)(=O)O

Specifications

Melting Point 300°C
Color Blue-White
Quantity 25 g
Formula Weight 297.33
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diaminodiphenylamine Sulfate Hydrate
EINECSNumber (3)-2717
TSCA No