missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diaminodicyclohexylmethane, mixture of stereoisomers, 98+%

Catalog No. AAL0959822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0959822 100 g
AAL0959814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0959822 Supplier Thermo Scientific Chemicals Supplier No. L0959822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1761-71-3 | C13H26N2 | 210.365 g/mol

4,4'-Diaminodicyclohexylmethane is used as epoxy curing agents. It is a versatile intermediate to produce leather, rubber products, plastics, dyes, and photo sensitive polymers. It is used in manufacturing diisocyanates and polyamides. It is also used as lubricant additive as and corrosion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Diaminodicyclohexylmethane is used as epoxy curing agents. It is a versatile intermediate to produce leather, rubber products, plastics, dyes, and photo sensitive polymers. It is used in manufacturing diisocyanates and polyamides. It is also used as lubricant additive as and corrosion inhibitor.

Solubility
Soluble in water. (5 g/L) at 20°C.

Notes
Incompatible materials are acids, acid chlorides, acid anhydrides, oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 1761-71-3
Molecular Formula C13H26N2
Molecular Weight (g/mol) 210.365
MDL Number MFCD00001496
InChI Key DZIHTWJGPDVSGE-UHFFFAOYSA-N
Synonym 4,4'-methylenedicyclohexanamine, 4,4'-diaminodicyclohexylmethane, 4,4'-methylenebis cyclohexylamine, cyclohexanamine, 4,4'-methylenebis, wandamin hm, bis 4-aminocyclohexyl methane, bis p-aminocyclohexyl methane, 4,4'-diaminodicyclohexyl methane, pacm 20, cyclohexylamine, 4,4'-methylenebis
PubChem CID 15660
IUPAC Name 4-[(4-aminocyclohexyl)methyl]cyclohexan-1-amine
SMILES C1CC(CCC1CC2CCC(CC2)N)N

Specifications

Melting Point ∼43°C
pH 11.5
Boiling Point 325°C to 327°C
Flash Point 159°C (318°F)
Odor Amine-like
Quantity 100 g
UN Number UN2928
Beilstein 2079179
Solubility Information Soluble in water. (5g/L) at 20°C.
Formula Weight 210.37
Percent Purity ≥98%
Chemical Name or Material 4,4'-Diaminodicyclohexylmethane, mixture of stereoisomers
Show More Show Less
Hazard Category H302-H314-H317-H335-H373
Hazard Statement GHS H Statement
H330-H314-H318-H302-H317
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: I
EINECSNumber 217-168-8
RTECSNumber GX1530000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only