missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Diacetylbiphenyl 99.0+%, TCI America™
$240.46 - $439.92
Chemical Identifiers
| CAS | 787-69-9 |
|---|---|
| Molecular Formula | C16H14O2 |
| Molecular Weight (g/mol) | 238.286 |
| MDL Number | MFCD00017248 |
| InChI Key | YSTSBXDVNKYPTR-UHFFFAOYSA-N |
| Synonym | 4,4'-diacetylbiphenyl, 4,4'-diacetyl biphenyl, 1-4-4-acetylphenyl phenyl ethanone, 4,4-diacetylbiphenyl, 1,1'-1,1'-biphenyl-4,4'-diyl diethanone, 1-4'-acetyl 1,1'-biphenyl-4-yl ethanone, ethanone, 1,1'-1,1'-biphenyl-4,4'-diylbis, 1-acetyl-4-4-acetylphenyl benzene, 1-4'-acetyl-1,1'-biphenyl-4-yl ethanone, diacetyl biphenyl |
| PubChem CID | 301558 |
| IUPAC Name | 1-[4-(4-acetylphenyl)phenyl]ethanone |
| SMILES | CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C |
Chemical Identifiers
| 787-69-9 | |
| 238.286 | |
| YSTSBXDVNKYPTR-UHFFFAOYSA-N | |
| 301558 | |
| CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C |
| C16H14O2 | |
| MFCD00017248 | |
| 4,4'-diacetylbiphenyl, 4,4'-diacetyl biphenyl, 1-4-4-acetylphenyl phenyl ethanone, 4,4-diacetylbiphenyl, 1,1'-1,1'-biphenyl-4,4'-diyl diethanone, 1-4'-acetyl 1,1'-biphenyl-4-yl ethanone, ethanone, 1,1'-1,1'-biphenyl-4,4'-diylbis, 1-acetyl-4-4-acetylphenyl benzene, 1-4'-acetyl-1,1'-biphenyl-4-yl ethanone, diacetyl biphenyl | |
| 1-[4-(4-acetylphenyl)phenyl]ethanone |
Specifications
| 787-69-9 | |
| Yellow | |
| MFCD00017248 | |
| 3077 | |
| YSTSBXDVNKYPTR-UHFFFAOYSA-N | |
| 1-[4-(4-acetylphenyl)phenyl]ethanone | |
| 301558 | |
| ≥99.0% (GC) | |
| 4,4′-Diacetylbiphenyl |
| 194°C | |
| C16H14O2 | |
| 10 g | |
| 4,4'-diacetylbiphenyl, 4,4'-diacetyl biphenyl, 1-4-4-acetylphenyl phenyl ethanone, 4,4-diacetylbiphenyl, 1,1'-1,1'-biphenyl-4,4'-diyl diethanone, 1-4'-acetyl 1,1'-biphenyl-4-yl ethanone, ethanone, 1,1'-1,1'-biphenyl-4,4'-diylbis, 1-acetyl-4-4-acetylphenyl benzene, 1-4'-acetyl-1,1'-biphenyl-4-yl ethanone, diacetyl biphenyl | |
| CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C | |
| 238.286 | |
| 238.29 | |
| Crystalline Powder |
Safety and Handling
TSCA : No