missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diacetylbiphenyl 99.0+%, TCI America™

Catalog No. D190210G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D190210G 10 g
D190225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D190210G Supplier TCI America Supplier No. D190210G

Chemical Identifiers

CAS 787-69-9
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD00017248
InChI Key YSTSBXDVNKYPTR-UHFFFAOYSA-N
Synonym 4,4'-diacetylbiphenyl, 4,4'-diacetyl biphenyl, 1-4-4-acetylphenyl phenyl ethanone, 4,4-diacetylbiphenyl, 1,1'-1,1'-biphenyl-4,4'-diyl diethanone, 1-4'-acetyl 1,1'-biphenyl-4-yl ethanone, ethanone, 1,1'-1,1'-biphenyl-4,4'-diylbis, 1-acetyl-4-4-acetylphenyl benzene, 1-4'-acetyl-1,1'-biphenyl-4-yl ethanone, diacetyl biphenyl
PubChem CID 301558
IUPAC Name 1-[4-(4-acetylphenyl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C

Specifications

Melting Point 194°C
Color Yellow
Quantity 10 g
UN Number 3077
Formula Weight 238.29
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diacetylbiphenyl