missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diacetylbiphenyl, 98%, Thermo Scientific™

Chemical Identifiers

CAS 787-69-9
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.28
InChI Key YSTSBXDVNKYPTR-UHFFFAOYSA-N
Synonym 4,4'-diacetylbiphenyl, 4,4'-diacetyl biphenyl, 1-4-4-acetylphenyl phenyl ethanone, 4,4-diacetylbiphenyl, 1,1'-1,1'-biphenyl-4,4'-diyl diethanone, 1-4'-acetyl 1,1'-biphenyl-4-yl ethanone, ethanone, 1,1'-1,1'-biphenyl-4,4'-diylbis, 1-acetyl-4-4-acetylphenyl benzene, 1-4'-acetyl-1,1'-biphenyl-4-yl ethanone, diacetyl biphenyl
PubChem CID 301558
IUPAC Name 1-[4-(4-acetylphenyl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC305440010
SDS View Documents
Thermo Scientific Chemicals
305440010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC305440050
SDS View Documents
Thermo Scientific Chemicals
305440050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

787-69-9
238.28
4,4'-diacetylbiphenyl, 4,4'-diacetyl biphenyl, 1-4-4-acetylphenyl phenyl ethanone, 4,4-diacetylbiphenyl, 1,1'-1,1'-biphenyl-4,4'-diyl diethanone, 1-4'-acetyl 1,1'-biphenyl-4-yl ethanone, ethanone, 1,1'-1,1'-biphenyl-4,4'-diylbis, 1-acetyl-4-4-acetylphenyl benzene, 1-4'-acetyl-1,1'-biphenyl-4-yl ethanone, diacetyl biphenyl
1-[4-(4-acetylphenyl)phenyl]ethanone
C16H14O2
YSTSBXDVNKYPTR-UHFFFAOYSA-N
301558
CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C

Specifications

787-69-9
100.0
97.5% min. (GC)
C16H14O2
1g
YSTSBXDVNKYPTR-UHFFFAOYSA-N
1-[4-(4-acetylphenyl)phenyl]ethanone
301558
98%
97.5
Authentic
Glass bottle
CH3COC6H4C6H4COCH3
4,4'-diacetylbiphenyl, 4,4'-diacetyl biphenyl, 1-4-4-acetylphenyl phenyl ethanone, 4,4-diacetylbiphenyl, 1,1'-1,1'-biphenyl-4,4'-diyl diethanone, 1-4'-acetyl 1,1'-biphenyl-4-yl ethanone, ethanone, 1,1'-1,1'-biphenyl-4,4'-diylbis, 1-acetyl-4-4-acetylphenyl benzene, 1-4'-acetyl-1,1'-biphenyl-4-yl ethanone, diacetyl biphenyl
CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C
238.28
238.28
4, 4'-Diacetylbiphenyl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Very toxic to aquatic life.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only