missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diacetylbiphenyl, 98%

Catalog No. AC305440050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC305440050 5 g Glass bottle
AC305440010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC305440050 Supplier Thermo Scientific Chemicals Supplier No. 305440050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 787-69-9 | C16H14O2 | 238.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 787-69-9
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.28
InChI Key YSTSBXDVNKYPTR-UHFFFAOYSA-N
Synonym 4,4'-diacetylbiphenyl, 4,4'-diacetyl biphenyl, 1-4-4-acetylphenyl phenyl ethanone, 4,4-diacetylbiphenyl, 1,1'-1,1'-biphenyl-4,4'-diyl diethanone, 1-4'-acetyl 1,1'-biphenyl-4-yl ethanone, ethanone, 1,1'-1,1'-biphenyl-4,4'-diylbis, 1-acetyl-4-4-acetylphenyl benzene, 1-4'-acetyl-1,1'-biphenyl-4-yl ethanone, diacetyl biphenyl
PubChem CID 301558
IUPAC Name 1-[4-(4-acetylphenyl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 192.0°C to 196.0°C
Color Colorless to Tan
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3COC6H4C6H4COCH3
Quantity 5 g
Formula Weight 238.28
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-Diacetylbiphenyl
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Very toxic to aquatic life.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

RUO – Research Use Only