missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diacetylbiphenyl, 98%

Catalog No. AAA1591106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1591106 5 g
AAA1591114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1591106 Supplier Thermo Scientific Chemicals Supplier No. A1591106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 787-69-9 | C16H14O2 | 238.286 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 787-69-9
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD00017248
InChI Key YSTSBXDVNKYPTR-UHFFFAOYSA-N
Synonym 4,4'-diacetylbiphenyl, 4,4'-diacetyl biphenyl, 1-4-4-acetylphenyl phenyl ethanone, 4,4-diacetylbiphenyl, 1,1'-1,1'-biphenyl-4,4'-diyl diethanone, 1-4'-acetyl 1,1'-biphenyl-4-yl ethanone, ethanone, 1,1'-1,1'-biphenyl-4,4'-diylbis, 1-acetyl-4-4-acetylphenyl benzene, 1-4'-acetyl-1,1'-biphenyl-4-yl ethanone, diacetyl biphenyl
PubChem CID 301558
IUPAC Name 1-[4-(4-acetylphenyl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)C

Specifications

Melting Point 190°C to 193°C
Quantity 5 g
Formula Weight 238.29
Percent Purity 98%
Chemical Name or Material 4,4'-Diacetylbiphenyl
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only