missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Di-tert-butylbiphenyl 98.0+%, TCI America™
$42.87 - $164.03
Chemical Identifiers
| CAS | 1625-91-8 |
|---|---|
| Molecular Formula | C20H26 |
| Molecular Weight (g/mol) | 266.428 |
| MDL Number | MFCD00008834 |
| InChI Key | CDKCEZNPAYWORX-UHFFFAOYSA-N |
| Synonym | 4,4'-di-tert-butylbiphenyl, 4,4'-di-tert-butyl-1,1'-biphenyl, 4,4'-di-t-butylbiphenyl, 1-tert-butyl-4-4-tert-butylphenyl benzene, 1,1'-biphenyl, 4,4'-bis 1,1-dimethylethyl, 1-tert-butyl-4-4-tert-butyl phenyl benzene, di-t-butylbiphenyl, pubchem9048, acmc-209st2, 4,4'-di-tertbutylbiphenyl |
| PubChem CID | 74195 |
| IUPAC Name | 1-tert-butyl-4-(4-tert-butylphenyl)benzene |
| SMILES | CC(C)(C)C1=CC=C(C=C1)C2=CC=C(C=C2)C(C)(C)C |
Chemical Identifiers
| 1625-91-8 | |
| 266.428 | |
| CDKCEZNPAYWORX-UHFFFAOYSA-N | |
| 74195 | |
| CC(C)(C)C1=CC=C(C=C1)C2=CC=C(C=C2)C(C)(C)C |
| C20H26 | |
| MFCD00008834 | |
| 4,4'-di-tert-butylbiphenyl, 4,4'-di-tert-butyl-1,1'-biphenyl, 4,4'-di-t-butylbiphenyl, 1-tert-butyl-4-4-tert-butylphenyl benzene, 1,1'-biphenyl, 4,4'-bis 1,1-dimethylethyl, 1-tert-butyl-4-4-tert-butyl phenyl benzene, di-t-butylbiphenyl, pubchem9048, acmc-209st2, 4,4'-di-tertbutylbiphenyl | |
| 1-tert-butyl-4-(4-tert-butylphenyl)benzene |
Specifications
| 1625-91-8 | |
| White-Yellow | |
| C20H26 | |
| 5 g | |
| CDKCEZNPAYWORX-UHFFFAOYSA-N | |
| 1-tert-butyl-4-(4-tert-butylphenyl)benzene | |
| 74195 | |
| ≥98.0% (GC) | |
| 4,4′-Di-tert-butylbiphenyl |
| 128°C | |
| 192°C | |
| MFCD00008834 | |
| 4,4'-di-tert-butylbiphenyl, 4,4'-di-tert-butyl-1,1'-biphenyl, 4,4'-di-t-butylbiphenyl, 1-tert-butyl-4-4-tert-butylphenyl benzene, 1,1'-biphenyl, 4,4'-bis 1,1-dimethylethyl, 1-tert-butyl-4-4-tert-butyl phenyl benzene, di-t-butylbiphenyl, pubchem9048, acmc-209st2, 4,4'-di-tertbutylbiphenyl | |
| CC(C)(C)C1=CC=C(C=C1)C2=CC=C(C=C2)C(C)(C)C | |
| 266.428 | |
| 266.43 | |
| Crystalline Powder |
Safety and Handling
TSCA : No