missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Di-tert-butylbiphenyl 98.0+%, TCI America™

Catalog No. D24505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24505G 5 g
D245025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24505G Supplier TCI America Supplier No. D24505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1625-91-8
Molecular Formula C20H26
Molecular Weight (g/mol) 266.428
MDL Number MFCD00008834
InChI Key CDKCEZNPAYWORX-UHFFFAOYSA-N
Synonym 4,4'-di-tert-butylbiphenyl, 4,4'-di-tert-butyl-1,1'-biphenyl, 4,4'-di-t-butylbiphenyl, 1-tert-butyl-4-4-tert-butylphenyl benzene, 1,1'-biphenyl, 4,4'-bis 1,1-dimethylethyl, 1-tert-butyl-4-4-tert-butyl phenyl benzene, di-t-butylbiphenyl, pubchem9048, acmc-209st2, 4,4'-di-tertbutylbiphenyl
PubChem CID 74195
IUPAC Name 1-tert-butyl-4-(4-tert-butylphenyl)benzene
SMILES CC(C)(C)C1=CC=C(C=C1)C2=CC=C(C=C2)C(C)(C)C

Specifications

Melting Point 128°C
Color White-Yellow
Boiling Point 192°C
Quantity 5 g
Formula Weight 266.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Di-tert-butylbiphenyl