missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Di-tert-butyl-2,2'-bipyridyl 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 161°C |
| Color | White |
| Quantity | 5 g |
| Synonym | 4,4′-Di-tert-butyl-2,2′-bipyridine |
| Formula Weight | 268.40 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,4′-Di-tert-butyl-2,2′-bipyridyl |
Safety and Handling
| TSCA | No |