missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Cyanophenyl)morpholine 98.0+%, TCI America™

Catalog No. C20085G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20085G Supplier TCI America Supplier No. C20085G
Only null left

Chemical Identifiers

CAS 10282-31-2
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
InChI Key ZSCUWVQXQDCSRV-UHFFFAOYSA-N
Synonym 4-morpholinobenzonitrile, 4-4-cyanophenyl morpholine, 4-morpholin-4-yl benzonitrile, 4-4-morpholinyl benzonitrile, 4-morpholin-4-yl-benzonitrile, benzonitrile, 4-4-morpholinyl, benzonitrile,4-4-morpholinyl, 4-morpholin-4-ylbenzenecarbonitrile, pubchem18708, acmc-1bqst
PubChem CID 394808
IUPAC Name 4-morpholin-4-ylbenzonitrile
SMILES C1COCCN1C2=CC=C(C=C2)C#N

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 188.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Cyanophenyl)morpholine