Learn More
4-[(4-chlorophenyl)sulfonyl]-3-methylthiophene-2-carboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 175137-65-2 |
|---|---|
| Molecular Formula | C12H9ClO4S2 |
| Molecular Weight (g/mol) | 316.77 |
| MDL Number | MFCD00067886 |
| InChI Key | WZQQSJVPCVCQKE-UHFFFAOYSA-N |
| Synonym | 4-4-chlorophenyl sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorobenzenesulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chloro-benzenesulfonyl-3-methyl-thiophene-2-carboxylic acid, maybridge1_004536, 4-4-chlorophenylsulfonyl-3-methylthiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 4-4-chlorophenyl sulfonyl-3-methyl, 4-4-chlorobenzene-1-sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorophenyl sulfonyl-3-methyl-thiophene-2-carboxylic acid |
| PubChem CID | 2778453 |
| IUPAC Name | 4-(4-chlorophenyl)sulfonyl-3-methylthiophene-2-carboxylic acid |
| SMILES | CC1=C(SC=C1S(=O)(=O)C2=CC=C(C=C2)Cl)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
KM01338DA
|
Thermo Scientific Chemicals
KM01338DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 175137-65-2 | |
| 316.77 | |
| WZQQSJVPCVCQKE-UHFFFAOYSA-N | |
| 2778453 | |
| CC1=C(SC=C1S(=O)(=O)C2=CC=C(C=C2)Cl)C(=O)O |
| C12H9ClO4S2 | |
| MFCD00067886 | |
| 4-4-chlorophenyl sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorobenzenesulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chloro-benzenesulfonyl-3-methyl-thiophene-2-carboxylic acid, maybridge1_004536, 4-4-chlorophenylsulfonyl-3-methylthiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 4-4-chlorophenyl sulfonyl-3-methyl, 4-4-chlorobenzene-1-sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorophenyl sulfonyl-3-methyl-thiophene-2-carboxylic acid | |
| 4-(4-chlorophenyl)sulfonyl-3-methylthiophene-2-carboxylic acid |
Specifications
| 175137-65-2 | |
| 97% | |
| C12H9ClO4S2 | |
| 1g | |
| WZQQSJVPCVCQKE-UHFFFAOYSA-N | |
| 4-(4-chlorophenyl)sulfonyl-3-methylthiophene-2-carboxylic acid | |
| 2778453 | |
| 97% |
| 97.0 | |
| Amber Glass Bottle | |
| MFCD00067886 | |
| 4-4-chlorophenyl sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorobenzenesulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chloro-benzenesulfonyl-3-methyl-thiophene-2-carboxylic acid, maybridge1_004536, 4-4-chlorophenylsulfonyl-3-methylthiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 4-4-chlorophenyl sulfonyl-3-methyl, 4-4-chlorobenzene-1-sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorophenyl sulfonyl-3-methyl-thiophene-2-carboxylic acid | |
| CC1=C(SC=C1S(=O)(=O)C2=CC=C(C=C2)Cl)C(=O)O | |
| 316.77 | |
| 316.78 | |
| 4-[(4-chlorophenyl)sulfonyl]-3-methylthiophene-2-carboxylic acid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.