missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorophenyl)-4-hydroxypiperidine, 99+%, crystalline

Catalog No. AC296840050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296840050 5 g Glass bottle
AC296840010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC296840050 Supplier Thermo Scientific Chemicals Supplier No. 296840050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39512-49-7 | C11H15Cl2NO | 248.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39512-49-7
Molecular Formula C11H15Cl2NO
Molecular Weight (g/mol) 248.15
MDL Number MFCD00006001
InChI Key PXAJMBHRLGKPQV-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl piperidin-4-ol, 4-4-chlorophenyl-4-hydroxypiperidine, 4-4-chlorophenyl-4-piperidinol, 4-chlorophenyl-4-hydroxylpiperidine, 4-p-chlorophenyl-4-hydroxypiperidine, 4-p-chlorophenyl piperidin-4-ol, 4-piperidinol, 4-4-chlorophenyl, haloperidol metabolite i, unii-und92fks0w, 4-hydroxy-4-4-chlorophenyl piperidine
PubChem CID 38282
IUPAC Name 4-(4-chlorophenyl)piperidin-4-ol
SMILES [H+].[Cl-].OC1(CCNCC1)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 99.0
Absorbance (1 w/v% soln. in DMF) (1 cm silica cell), (1 w/v% soln. in DMF) (1 cm silica cell) at 0nm, 0.05 max. at 440nm
CAS Max % 100.0
Melting Point 139°C to 141°C
Color Cream-White to White
Loss on Drying 0.5% max. (1 g, 105°C, 2 hrs, 760mm Hg)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: 340mg/l (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,dmf,,acetone and dichloromethane
Formula Weight 211.69
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Chlorophenyl)-4-hydroxypiperidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 254-479-8

RUO – Research Use Only