missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorophenoxy)piperidine,98%

Catalog No. p-7033776
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50944MD 250 mg
AAH5094403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50944MD Supplier Thermo Scientific Chemicals Supplier No. H50944MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 97839-99-1 | C11H14ClNO | 211.69 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 97839-99-1
Molecular Formula C11H14ClNO
Molecular Weight (g/mol) 211.69
MDL Number MFCD04113544
InChI Key VOQMPZXAFLPTMM-UHFFFAOYSA-N
PubChem CID 5199889
IUPAC Name 4-(4-chlorophenoxy)piperidine
SMILES ClC1=CC=C(OC2CCNCC2)C=C1

Specifications

Quantity 250 mg
Synonym 4-4-chlorophenoxy piperidine, 4-4-chloro-phenoxy-piperidine, piperidine, 4-4-chlorophenoxy, acmc-20amwb, 4-p-chlorophenoxy piperidine, 4-4-chlorophenyl oxy piperidine, 4-chloro-1-4-piperidyloxy benzene, piperidine,4-4-chlorophenoxy, piperidine, 4-4-chlorophenoxy-, hydrochloride
Solubility Information Slightly soluble in water.
Formula Weight 211.69
Percent Purity 98%
Chemical Name or Material 4-(4-Chlorophenoxy)piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only