missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorobenzyl)pyridine 95.0+%, TCI America™

Catalog No. C105425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C105425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C105425G Supplier TCI America Supplier No. C105425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4409-11-4
Molecular Formula C12H10ClN
Molecular Weight (g/mol) 203.67
MDL Number MFCD00006444
InChI Key OHKBVLWPESSWKC-UHFFFAOYSA-N
Synonym 4-4-chlorobenzyl pyridine, 4-4-chlorophenyl methyl pyridine, 4-4'-chlorobenzyl pyridine, 4-4-chloro-benzyl-pyridine, 4-para-chlorobenzyl-pyridine, pyridine, 4-4-chlorophenyl methyl, acmc-2097fd, 4-p-chlorobenzyl pyridine
PubChem CID 78124
IUPAC Name 4-[(4-chlorophenyl)methyl]pyridine
SMILES C1=CC(=CC=C1CC2=CC=NC=C2)Cl

Specifications

Color Yellow
Quantity 25 g
Physical Form Crystal at 20°C
Formula Weight 203.67
Percent Purity ≥95.0% (GC)
Chemical Name or Material 4-(4-Chlorobenzyl)pyridine
RTECSNumber US7204100
TSCA No