missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Carbonyldimorpholine 98.0+%, TCI America™

Catalog No. C29621G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29621G 1 g
C29625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29621G Supplier TCI America Supplier No. C29621G
Only null left

Chemical Identifiers

CAS 38952-62-4
Molecular Formula C9H16N2O3
Molecular Weight (g/mol) 200.24
MDL Number MFCD00593814
InChI Key QONGECDDDTYBGS-UHFFFAOYSA-N
PubChem CID 655428
IUPAC Name 4-(morpholine-4-carbonyl)morpholine
SMILES O=C(N1CCOCC1)N1CCOCC1

Specifications

Melting Point 144°C
Color White
Quantity 1 g
Formula Weight 200.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Carbonyldimorpholine