missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Bromophenyl)dibenzofuran 98.0+%, TCI America™
$154.02 - $610.28
Chemical Identifiers
| CAS | 955959-84-9 |
|---|---|
| Molecular Formula | C18H11BrO |
| Molecular Weight (g/mol) | 323.189 |
| MDL Number | MFCD19441306 |
| InChI Key | RIPZSADLUWTEFQ-UHFFFAOYSA-N |
| PubChem CID | 59022098 |
| IUPAC Name | 4-(4-bromophenyl)dibenzofuran |
| SMILES | C1=CC=C2C(=C1)C3=C(O2)C(=CC=C3)C4=CC=C(C=C4)Br |
Chemical Identifiers
| 955959-84-9 | |
| 323.189 | |
| RIPZSADLUWTEFQ-UHFFFAOYSA-N | |
| 4-(4-bromophenyl)dibenzofuran |
| C18H11BrO | |
| MFCD19441306 | |
| 59022098 | |
| C1=CC=C2C(=C1)C3=C(O2)C(=CC=C3)C4=CC=C(C=C4)Br |
Specifications
| 955959-84-9 | |
| White | |
| MFCD19441306 | |
| RIPZSADLUWTEFQ-UHFFFAOYSA-N | |
| 4-(4-bromophenyl)dibenzofuran | |
| 59022098 | |
| ≥98.0% (GC) | |
| 4-(4-Bromophenyl)dibenzofuran |
| 101°C | |
| C18H11BrO | |
| 1 g | |
| C1=CC=C2C(=C1)C3=C(O2)C(=CC=C3)C4=CC=C(C=C4)Br | |
| 323.189 | |
| 323.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No