missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromophenyl)dibenzofuran 98.0+%, TCI America™

Catalog No. B48841G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48841G 1 g
B48845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48841G Supplier TCI America Supplier No. B48841G

Chemical Identifiers

CAS 955959-84-9
Molecular Formula C18H11BrO
Molecular Weight (g/mol) 323.189
MDL Number MFCD19441306
InChI Key RIPZSADLUWTEFQ-UHFFFAOYSA-N
PubChem CID 59022098
IUPAC Name 4-(4-bromophenyl)dibenzofuran
SMILES C1=CC=C2C(=C1)C3=C(O2)C(=CC=C3)C4=CC=C(C=C4)Br

Specifications

Melting Point 101°C
Color White
Quantity 1 g
Formula Weight 323.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Bromophenyl)dibenzofuran