missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Bromophenoxy)benzaldehyde 98.0+%, TCI America™
$171.49 - $546.17
Chemical Identifiers
| CAS | 69240-56-8 |
|---|---|
| Molecular Formula | C13H9BrO2 |
| Molecular Weight (g/mol) | 277.117 |
| MDL Number | MFCD08276811 |
| InChI Key | WMDDJOBNAOVSLP-UHFFFAOYSA-N |
| Synonym | 4-4-bromophenoxy benzaldehyde, 4-4-bromo-phenoxy benzaldehyde, 4-4-bromo-phenoxy-benzaldehyde, acmc-20amwk |
| PubChem CID | 10636240 |
| IUPAC Name | 4-(4-bromophenoxy)benzaldehyde |
| SMILES | C1=CC(=CC=C1C=O)OC2=CC=C(C=C2)Br |
Chemical Identifiers
| 69240-56-8 | |
| 277.117 | |
| WMDDJOBNAOVSLP-UHFFFAOYSA-N | |
| 10636240 | |
| C1=CC(=CC=C1C=O)OC2=CC=C(C=C2)Br |
| C13H9BrO2 | |
| MFCD08276811 | |
| 4-4-bromophenoxy benzaldehyde, 4-4-bromo-phenoxy benzaldehyde, 4-4-bromo-phenoxy-benzaldehyde, acmc-20amwk | |
| 4-(4-bromophenoxy)benzaldehyde |
Specifications
| 69240-56-8 | |
| White-Yellow | |
| C13H9BrO2 | |
| 1 g | |
| WMDDJOBNAOVSLP-UHFFFAOYSA-N | |
| 4-(4-bromophenoxy)benzaldehyde | |
| 10636240 | |
| ≥98.0% (GC) | |
| 4-(4-Bromophenoxy)benzaldehyde |
| 71°C | |
| 189°C | |
| MFCD08276811 | |
| 4-4-bromophenoxy benzaldehyde, 4-4-bromo-phenoxy benzaldehyde, 4-4-bromo-phenoxy-benzaldehyde, acmc-20amwk | |
| C1=CC(=CC=C1C=O)OC2=CC=C(C=C2)Br | |
| 277.117 | |
| 277.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No