missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromophenoxy)benzaldehyde 98.0+%, TCI America™

Catalog No. B49001G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49001G 1 g
B49005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49001G Supplier TCI America Supplier No. B49001G

Chemical Identifiers

CAS 69240-56-8
Molecular Formula C13H9BrO2
Molecular Weight (g/mol) 277.117
MDL Number MFCD08276811
InChI Key WMDDJOBNAOVSLP-UHFFFAOYSA-N
Synonym 4-4-bromophenoxy benzaldehyde, 4-4-bromo-phenoxy benzaldehyde, 4-4-bromo-phenoxy-benzaldehyde, acmc-20amwk
PubChem CID 10636240
IUPAC Name 4-(4-bromophenoxy)benzaldehyde
SMILES C1=CC(=CC=C1C=O)OC2=CC=C(C=C2)Br

Specifications

Melting Point 71°C
Color White-Yellow
Boiling Point 189°C
Quantity 1 g
Formula Weight 277.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Bromophenoxy)benzaldehyde