Learn More
4-(4-Bromophenoxy)benzaldehyde, 97%
CAS: 69240-56-8 | C13H9BrO2 | 277.117 g/mol
$133.11 - $443.10
Chemical Identifiers
| CAS | 69240-56-8 |
|---|---|
| Molecular Formula | C13H9BrO2 |
| Molecular Weight (g/mol) | 277.117 |
| MDL Number | MFCD08276811 |
| InChI Key | WMDDJOBNAOVSLP-UHFFFAOYSA-N |
| Synonym | 4-4-bromophenoxy benzaldehyde, 4-4-bromo-phenoxy benzaldehyde, 4-4-bromo-phenoxy-benzaldehyde, acmc-20amwk |
| PubChem CID | 10636240 |
| IUPAC Name | 4-(4-bromophenoxy)benzaldehyde |
| SMILES | C1=CC(=CC=C1C=O)OC2=CC=C(C=C2)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3170203
|
Thermo Scientific Chemicals
H3170203 |
1 g |
Each for $133.11
|
|
|||||
|
AAH3170206
|
Thermo Scientific Chemicals
H3170206 |
5 g |
Each for $443.10
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 69240-56-8 | |
| 277.117 | |
| WMDDJOBNAOVSLP-UHFFFAOYSA-N | |
| 10636240 | |
| C1=CC(=CC=C1C=O)OC2=CC=C(C=C2)Br |
| C13H9BrO2 | |
| MFCD08276811 | |
| 4-4-bromophenoxy benzaldehyde, 4-4-bromo-phenoxy benzaldehyde, 4-4-bromo-phenoxy-benzaldehyde, acmc-20amwk | |
| 4-(4-bromophenoxy)benzaldehyde |
Specifications
| 69240-56-8 | |
| C13H9BrO2 | |
| 1 g | |
| Air sensitive | |
| WMDDJOBNAOVSLP-UHFFFAOYSA-N | |
| 4-(4-bromophenoxy)benzaldehyde | |
| 10636240 | |
| 97% |
| 69°C to 73°C | |
| MFCD08276811 | |
| UN3077 | |
| 4-4-bromophenoxy benzaldehyde, 4-4-bromo-phenoxy benzaldehyde, 4-4-bromo-phenoxy-benzaldehyde, acmc-20amwk | |
| C1=CC(=CC=C1C=O)OC2=CC=C(C=C2)Br | |
| 277.117 | |
| 277.12 | |
| 4-(4-Bromophenoxy)benzaldehyde |
Safety and Handling
GHS H Statement
H318-H317
Causes serious eye damage.
May cause an allergic skin reaction.
P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P501c
H317-H318
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only