missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromophenoxy)benzaldehyde, 97%

Catalog No. AAH3170203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3170203 1 g
AAH3170206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3170203 Supplier Thermo Scientific Chemicals Supplier No. H3170203

CAS: 69240-56-8 | C13H9BrO2 | 277.117 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69240-56-8
Molecular Formula C13H9BrO2
Molecular Weight (g/mol) 277.117
MDL Number MFCD08276811
InChI Key WMDDJOBNAOVSLP-UHFFFAOYSA-N
Synonym 4-4-bromophenoxy benzaldehyde, 4-4-bromo-phenoxy benzaldehyde, 4-4-bromo-phenoxy-benzaldehyde, acmc-20amwk
PubChem CID 10636240
IUPAC Name 4-(4-bromophenoxy)benzaldehyde
SMILES C1=CC(=CC=C1C=O)OC2=CC=C(C=C2)Br

Specifications

Melting Point 69°C to 73°C
Quantity 1 g
UN Number UN3077
Sensitivity Air sensitive
Formula Weight 277.12
Percent Purity 97%
Chemical Name or Material 4-(4-Bromophenoxy)benzaldehyde
Hazard Category H317-H318
Hazard Statement GHS H Statement
H318-H317
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only