missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester, 95%

Catalog No. AAH5174306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5174306 5 g
AAH5174303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5174306 Supplier Thermo Scientific Chemicals Supplier No. H5174306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 936694-19-8 | C22H35BN2O4 | 402.342 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 936694-19-8
Molecular Formula C22H35BN2O4
Molecular Weight (g/mol) 402.342
MDL Number MFCD16294502
InChI Key AAEYFMAHSYKHGD-UHFFFAOYSA-N
Synonym tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperazine-1-carboxylate, tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperazine-1-carboxylate, 4-4-boc-piperazinomethyl phenylboronic acid, pinacol ester, 4-4-boc-1-piperazinylmethyl benzeneboronic acid pinacol ester, amtb122, 4-4-boc-piperazinemethyl phenylboronic acid pinacol ester, 4-4-boc-piperazine methyl phenylboronic acid pinacol ester, tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzyl piperazine-1-carboxylate
PubChem CID 53216820
IUPAC Name tert-butyl 4-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperazine-1-carboxylate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN3CCN(CC3)C(=O)OC(C)(C)C

Specifications

Quantity 5 g
Beilstein 18669672
Formula Weight 402.34
Percent Purity 95%
Chemical Name or Material 4-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only