missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(methoxymethyl)biphenyl 96.0+%, TCI America™

Catalog No. b304625g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Catalog No. Quantity
B304625G 25 g
1 options

Catalog No. B304625G

Supplier: TCI America B304625G

Only null left

Chemical Identifiers

CAS 3753-18-2
Molecular Formula C16H18O2
Molecular Weight (g/mol) 242.318
MDL Number MFCD03840532
InChI Key MODAACUAXYPNJH-UHFFFAOYSA-N
PubChem CID 1519430
IUPAC Name 1-(methoxymethyl)-4-[4-(methoxymethyl)phenyl]benzene
SMILES COCC1=CC=C(C=C1)C2=CC=C(C=C2)COC

Specifications

Melting Point 55°C
Color White-Yellow
Boiling Point 350°C
Quantity 25 g
Formula Weight 242.32
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(methoxymethyl)biphenyl
EINECSNumber (4)-1777
TSCA No