missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(hexyloxy)-3-methylazobenzene 95.0+%, TCI America™

Catalog No. B45965G
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45965G 5 g
B4596500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45965G Supplier TCI America Supplier No. B45965G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1440509-03-4
Molecular Formula C25H36N2O2
Molecular Weight (g/mol) 396.575
InChI Key ULSNKXLZLMMAQJ-UHFFFAOYSA-N
PubChem CID 102341608
IUPAC Name (4-hexoxy-3-methylphenyl)-(4-hexoxyphenyl)diazene
SMILES CCCCCCOC1=CC=C(C=C1)N=NC2=CC(=C(C=C2)OCCCCCC)C

Specifications

Quantity 5 g
Formula Weight 396.58
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(hexyloxy)-3-methylazobenzene