missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(dodecyloxy)-3-methylazobenzene 93.0+%, TCI America™

$285.21 - $2126.89

Chemical Identifiers

CAS 1440509-01-2
Molecular Formula C37H60N2O2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B4598500MG
View Documents
TCI America
B4598500MG
500 mg
Each for $285.21
Only null left
 
B45985G
View Documents
TCI America
B45985G
5 g
Each for $2,126.89
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1440509-01-2
C37H60N2O2

Specifications

1440509-01-2
500 mg
≥93.0% (HPLC)
4,4′-Bis(dodecyloxy)-3-methylazobenzene
C37H60N2O2
564.90
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'