missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(dimethylamino)benzophenone, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 164491000

 View more versions of this product

Catalog No. AC164491000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4, 4'-Bis(dimethylamino)benzophenone
90-94-8
100.0
220°C
97.5% min. (HPLC)
C17H20N2O
MFCD00008312
14, 89
15, 6526
Solubility in water: 400mg/l (20°C). Other solubilities: soluble in alcohol,warm benzene and ether
CN(C)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(C)C
268.36
CHEBI:82347
98%
98%
97.5
>360°C
Authentic
Glass bottle
[(CH3)2NC6H4]2CO
100g
02,285
michler's ketone, 4,4'-bis dimethylamino benzophenone, michler ketone, bis 4-dimethylamino phenyl methanone, p,p'-michler's ketone, michlers ketone, methanone, bis 4-dimethylamino phenyl, tetramethyldiaminobenzophenone, p,p'-bis dimethylamino benzophenone
VVBLNCFGVYUYGU-UHFFFAOYSA-N
bis[4-(dimethylamino)phenyl]methanone
7031
268.35

Chemical Identifiers

90-94-8
268.36
VVBLNCFGVYUYGU-UHFFFAOYSA-N
7031
bis[4-(dimethylamino)phenyl]methanone
C17H20N2O
MFCD00008312
michler's ketone, 4,4'-bis dimethylamino benzophenone, michler ketone, bis 4-dimethylamino phenyl methanone, p,p'-michler's ketone, michlers ketone, methanone, bis 4-dimethylamino phenyl, tetramethyldiaminobenzophenone, p,p'-bis dimethylamino benzophenone
CHEBI:82347
CN(C)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Suspected of causing genetic defects.
May cause cancer.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-027-5

RTECSNumber : DJ0250000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only