missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(dimethylamino)benzil 99.0+%, TCI America™

Catalog No. p-7118925
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B12125G 5 g
B121225G 25 g
2 options

Catalog No. B12125G

Supplier: TCI America B12125G

Only null left

Chemical Identifiers

CAS 17078-27-2
Molecular Formula C18H20N2O2
Molecular Weight (g/mol) 296.37
MDL Number MFCD00051389
InChI Key AVFUVYIDYFXFSX-UHFFFAOYSA-N
Synonym 4,4'-bis dimethylamino benzil, 1,2-bis 4-dimethylamino phenyl ethane-1,2-dione, ethandione, bis 4-dimethylamino phenyl, bis 4-dimethylamino phenyl ethane-1,2-dione, acmc-209e1j, benzil-based compound, 35, bis n,n-dimethylamino benzil, 4,4/'-bis dimethylamino benzil, bis 4-dimethylamino phenyl diketone
PubChem CID 222570
IUPAC Name 1,2-bis[4-(dimethylamino)phenyl]ethane-1,2-dione
SMILES CN(C)C1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)N(C)C

Specifications

Melting Point 203°C
Quantity 5 g
Formula Weight 296.37
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(dimethylamino)benzil