missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(9H-carbazol-9-yl)biphenyl 98.0+%, TCI America™

Catalog No. p-7119735
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27131G 1 g
B27135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27131G Supplier TCI America Supplier No. B27131G
Only null left

Chemical Identifiers

CAS 58328-31-7
Molecular Formula C36H24N2
Molecular Weight (g/mol) 484.602
MDL Number MFCD00093417
InChI Key VFUDMQLBKNMONU-UHFFFAOYSA-N
PubChem CID 11248716
IUPAC Name 9-[4-(4-carbazol-9-ylphenyl)phenyl]carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=C(C=C4)C5=CC=C(C=C5)N6C7=CC=CC=C7C8=CC=CC=C86

Specifications

Melting Point 283°C
Color White-Yellow
Quantity 1 g
Synonym CBP, 4,4′C-Di(9H-carbazol-9-yl)-1,1′C-biphenyl
Formula Weight 484.60
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(9H-carbazol-9-yl)biphenyl