missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(5-methyl-2-benzoxazolyl)stilbene (purified by sublimation) 96.0+%, TCI America™

Catalog No. B26761G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B26761G Supplier TCI America Supplier No. B26761G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2397-00-4
Molecular Formula C30H22N2O2
Molecular Weight (g/mol) 442.518
MDL Number MFCD00191316
InChI Key OKEZAUMKBWTTCR-AATRIKPKSA-N
PubChem CID 6436901
IUPAC Name 5-methyl-2-[4-[(E)-2-[4-(5-methyl-1,3-benzoxazol-2-yl)phenyl]ethenyl]phenyl]-1,3-benzoxazole
SMILES CC1=CC2=C(C=C1)OC(=N2)C3=CC=C(C=C3)C=CC4=CC=C(C=C4)C5=NC6=C(O5)C=CC(=C6)C

Specifications

Quantity 1 g
Formula Weight 442.52
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(5-methyl-2-benzoxazolyl)stilbene (purified by sublimation)
EINECSNumber (5)-0169
TSCA Yes