missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(4,6-diphenyl-1,3,5-triazin-2-yl)biphenyl (This product is unavailable in the U.S.) 95.0+%, TCI America™

Catalog No. p-7123879
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4977200MG 200 mg
B49771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4977200MG Supplier TCI America Supplier No. B4977200MG

Chemical Identifiers

CAS 266349-83-1
Molecular Formula C42H28N6
Molecular Weight (g/mol) 616.73
MDL Number MFCD28291963
InChI Key UICMBMCOVLMLIE-UHFFFAOYSA-N
PubChem CID 22598582
IUPAC Name 2-[4'-(4,6-diphenyl-1,3,5-triazin-2-yl)-[1,1'-biphenyl]-4-yl]-4,6-diphenyl-1,3,5-triazine
SMILES C1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 200 mg
Formula Weight 616.73
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(4,6-diphenyl-1,3,5-triazin-2-yl)biphenyl (This product is unavailable in the U.S.)