missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl 98.0+%, TCI America™
$111.66 - $390.80
Chemical Identifiers
| CAS | 207611-87-8 |
|---|---|
| Molecular Formula | C24H32B2O4 |
| Molecular Weight (g/mol) | 406.14 |
| MDL Number | MFCD08669545 |
| InChI Key | INNVTCAYPFZCAL-UHFFFAOYSA-N |
| Synonym | 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl, 4,4'-biphenyldiboronic acid dipinacol ester, 4,4'-biphenyldiboronic acid pinacol ester, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl biphenyl, biphenyl-4,4'-diboronic acid bis pinacol ester, 4,4'-biphenyldiboronic acid bis pinacol ester, 4,4,5,5-tetramethyl-2-4'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl-4-yl-1,3,2-dioxaborolane, amtb257, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl biphenyl |
| PubChem CID | 10525359 |
| IUPAC Name | 4,4,5,5-tetramethyl-2-[4'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-yl]-1,3,2-dioxaborolane |
| SMILES | CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 |
Chemical Identifiers
| 207611-87-8 | |
| 406.14 | |
| INNVTCAYPFZCAL-UHFFFAOYSA-N | |
| 10525359 | |
| CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 |
| C24H32B2O4 | |
| MFCD08669545 | |
| 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl, 4,4'-biphenyldiboronic acid dipinacol ester, 4,4'-biphenyldiboronic acid pinacol ester, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl biphenyl, biphenyl-4,4'-diboronic acid bis pinacol ester, 4,4'-biphenyldiboronic acid bis pinacol ester, 4,4,5,5-tetramethyl-2-4'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl-4-yl-1,3,2-dioxaborolane, amtb257, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl biphenyl | |
| 4,4,5,5-tetramethyl-2-[4'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-yl]-1,3,2-dioxaborolane |
Specifications
| 207611-87-8 | |
| White | |
| MFCD08669545 | |
| 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl, 4,4'-biphenyldiboronic acid dipinacol ester, 4,4'-biphenyldiboronic acid pinacol ester, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl biphenyl, biphenyl-4,4'-diboronic acid bis pinacol ester, 4,4'-biphenyldiboronic acid bis pinacol ester, 4,4,5,5-tetramethyl-2-4'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl-4-yl-1,3,2-dioxaborolane, amtb257, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl biphenyl | |
| CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 | |
| 406.14 | |
| 406.14 | |
| Crystalline Powder |
| 262°C | |
| C24H32B2O4 | |
| 1 g | |
| INNVTCAYPFZCAL-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-2-[4'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-yl]-1,3,2-dioxaborolane | |
| 10525359 | |
| ≥98.0% (GC,T) | |
| 4,4′-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl |
Safety and Handling
TSCA : No